Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tropodithietic Acid (from Roseobacter gallaeciensis) ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

BVT-0152-M001 BVT-0152-M005
102513-564EA 131.54 USD InStock
102513-564 102513-566
Tropodithietic Acid (from Roseobacter gallaeciensis) ≥98% (by HPLC)
Tropodithietic Acid
Tropodithietic Acid is a bacterial signal substance.

  • Antibiotic
  • Shows anti-tumor activity
  • Cytotoxic against selected mammalian neuronal cell lines

Antibiotic. Isomeric to thiotropocin. Antibacterial. Active against Gram-positive and Gram-negative bacteria. Antifungal and anti-nematodical. Cytotoxic against selected mammalian neuronal cell lines.

Orange solid

Stable for at least one year after receipt when stored at -20°C. Store solutions at -20°C in the dark.

Soluble in DMSO, methanol or acetone.


Formula: C₈H₄O₃S₂
MW: 212.3 g/mol
Storage Temperature: Freezer
CAS Number: 750590-18-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty