Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nargenicin A1 (from Actinomyces sp. Gö301) ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

BVT-0204-M001 BVT-0204-M005
102513-604EA 180 USD InStock
102513-604 102513-606
Nargenicin A1 (from Actinomyces sp. Gö301) ≥98% (by HPLC)
Nargenicin A1
Nargenicin A1 is an antibiotic against gram-positive bacteria.

  • Inhibits cell proliferation
  • More effective against multi-resistant strains (MRSA)
  • Displays low cytotoxicity

Antibiotic against Gram-positive bacteria, particularly Staphylococcus and Clostridia. More effective against multi-resistant strains (MRSA) than erythromycin and vancomycin. Inhibits cell proliferation and induces HL-60 cell differentiation in combination with 1,25-Dihydroxyvitamin-D3 and ATRA. Displays low cytotoxicity compared to erythromycin and spiramycin.

Off-white powder

After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.

Soluble in DMSO or methanol.


Formula: C₂₈H₃₇NO₈
MW: 515.6 g/mol
Storage Temperature: Freezer
CAS Number: 70695-02-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty