Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thaxtomin A (from Streptomyces bottropensis Gö-Dra 17) ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

BVT-0206-M001 BVT-0206-M005
102513-608EA 249.23 USD InStock
102513-608 102513-610
Thaxtomin A (from Streptomyces bottropensis Gö-Dra 17) ≥98% (by HPLC)
Thaxtomin A
Thaxtomin A is a Diketopiperazine antibiotic.

  • Phytotoxin
  • Herbicide
  • Induces common scab disease of potato

Phytotoxin. Herbicide. Natural cellulose synthesis inhibitor. Plant cell necrosis inducer.

Yellow powder

After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.

Soluble in DMSO or dimethyl formamide; partially soluble in methanol or 100% ethanol; poorly soluble in water.


Formula: C₂₂H₂₂N₄O₆
MW: 438.4 g/mol
Storage Temperature: Freezer
CAS Number: 122380-18-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty