Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Agistatin B ≥98%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

BVT-0223-M001 BVT-0223-M005
102513-616EA 180 USD InStock
102513-616 102513-618
Agistatin B ≥98%
Agistatin B
Tricyclic analog of agistatin A. Cholesterol biosynthesis inhibitor.


Formula: C₁₁H₁₈O₄
MW: 214.3 g/mol
Storage Temperature: Freezer
CAS Number: 144096-46-8
Order Now
Specification Test Results
Source/Host Isolated from fungus A 6239.
Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in methanol, DMSO or acetone.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty