Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Xanthomegnin ≥97%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

BVT-0365-C500 BVT-0365-M001
102513-804EA 180 USD InStock
102513-804 102513-806
Xanthomegnin ≥97%
Xanthomegnin
Genotoxic mycotoxin. Antitumor compound. Potent iNOS (NOSII) inhibitor. Interferes with cellular respiratory processes.


Formula: C₃₀H₂₂O₁₂
MW: 574.5 g/mol
Storage Temperature: Freezer
CAS Number: 1685-91-2
Order Now
Specification Test Results
Source/Host Isolated from Penicillium citreo-viride.
Purity 97% (HPLC)
Appearance/Color Orange crystalline solid.
Solubility Soluble in chloroform or DMSO; poorly soluble in methanol or water.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty