HiTrap™ MabSelect SuRe™ Columns
Supplier: GE Healthcare - Life Sciences
11-0034-93 11-0034-94 11-0034-95 29-0491-04
95055-926PK 1422.66 USD InStock
95055-926 95056-024 95055-842 89501-386
HiTrap™ MabSelect SuRe™ Columns
Chromatography Columns
HiTrap MabSelect SuRe columns are prepacked with MabSelect SuRe, a BioProcess medium for purification of monoclonal antibodies from large sample volumes.
  • Withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH
  • Novel ligand design gives enhanced protease resistance resulting in lower ligand leakage
  • Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
  • Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs
The novel alkali-tolerant recombinant Protein A ligand allow cleaning-in-place (CIP).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty