West Condenser, Glassco
Supplier: Glassco
182.470.00 182.470.01 182.470.02 182.470.02A 182.470.02B 182.470.03 182.470.06
89426-654EA 88.04 USD InStock
89426-654 89426-658 89426-660 89426-664 89426-666 89426-668 89426-670
West Condenser, Glassco
Condensers
Condensers feature narrow annular space for fast coolant speed, thus ensuring high cooling efficiency.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty