T-Bore Glass Stopcock, High Vacuum, Chemglass
Supplier: Chemglass
CG-464-01 CG-464-02 CG-464-03
80074-286EA 188.97 USD InStock
80074-286 80074-288 80074-290
T-Bore Glass Stopcock, High Vacuum, Chemglass
Stopcocks
Solid glass high vacuum stopcock with the new Chemglass improved constant pressure retention device
This unique retaining system incorporates a glass plug having an extended snelling which accepts a stainless steel spring and clip. The spring keeps constant tension on the plug at all times and is easily removed and attached. Solid glass plug has a "T" shaped bore so that any two or all three arms can be connected at one time. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug. Each stopcock is supplied complete with a stainless steel spring and clip.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty