Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carbenicillin disodium salt USP


MBPC-2202 MBPC-2201
101384-990EA 547.61 USD InStock
101384-990 101384-988
Carbenicillin disodium salt USP
Carbenicillin disodium salt
Carbenicillin Disodium Salt is a penicillin derivative that interferes with cell wall synthesis in gram negative and positive bacteria used for ampicillin-resistant plasmids

Carbenicillin maintains stability after being autoclaved.

Certificaciones: USP Grade


Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty