Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gentamicin sulphate USP

Supplier: A.W. SCHÜRR GmbH

00149-1KG 00149-250G 00149-100G 00149-5G 00149-25G
102562-124EA 2440.77 USD InStock
102562-124 102586-426 102606-330 102614-744 102614-746
Gentamicin sulphate USP
Gentamicin sulphate
White to off-white powder

  • Optical rotation: +107° to +121°
  • Potency: ≥ 590 µg/mg (Anhydrous basis)


Melting Pt: 218…237 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270181
CAS Number: 1405-41-0
Merck Index: 13,04403
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty