Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-chloro-5-nitropyridine

Supplier: A.W. SCHÜRR GmbH

28117-1G 28117-5G
102628-958EA 126.03 USD InStock
102628-958 102628-960
2-Amino-3-chloro-5-nitropyridine
2-Amino-3-chloro-5-nitropyridine


Formula: C₅H₄ClN₃O₂
MW: 173.5600 g/mol
MDL Number: MFCD00160299
CAS Number: 22353-35-1
Order Now
Specification Test Results
Molecular Formula C5H4ClN3O2
Molecular Weight 173.56
MDL Number MFCD00160299
Storage Temperature Store at 0-8°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty