RNase A, Protease-Free, Highly Purified, Bovine Pancreas, MilliporeSigma
Supplier: EMD MILLIPORE (BIOSCIENCES)
556746-50KU 556746-10KU
80602-646EA 633.29 USD InStock
80602-646 80602-644
RNase A, Protease-Free, Highly Purified, Bovine Pancreas, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
Ribonuclease A, Protease-Free is a chromatographically purified, pyrimidine-specific endoribonuclease that acts on single-stranded RNA
For selective degradation of RNA.
One unit is defined as the amount of enzyme that will catalyze the hydrolysis of RNA to yield a first-order velocity constant equal to 1 at 25°C, pH 5.0.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty