Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Water, Cell Culture Grade pyrogen-, endotoxin-free, sterile


CCPW-1000-12 CCPW-2000-6
100751-988CS 352.84 USD InStock
100751-988 100751-992
Water, Cell Culture Grade pyrogen-, endotoxin-free, sterile
Water
Optimally prepared for adherent/suspension cell culture applications such as media and buffer preparation, water systems, incubators and for the preparation of reagents for chemical analysis or synthesis. Our cell culture water eliminates endotoxin concerns as it has no detectable Endotoxins (below the detection threshold of <0.005EU/mL) and contains no detectable gram-negative bacteria.
The water is prepared by distillation, 0.1 micron sterile filtration and steam sterilization directly in the dispensed bottles.

Growcells provides the highest quality of Nuclease Free water for critical applications. Water is purified, tested and certified to be devoid of any activiy in DNAse, RNAse, and Protease assays. Available in convenient volumes and packaging to fit all applications requiring biologically inactive water.
Polycarbonate bottle packaging is re-usable and autoclavable.


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098
Order Now
Specification Test Results
Sterile Filtered Yes (0.10 μM)
Storage RT
Shipping Ground
Shelf Life 3 Years (from Date of Manufacture)
Sterility SAL 10-6
Endotoxin <0.005eu/ml
Activity None Detected

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty