Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vinyltriethoxysilane for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Warning

M-Clarity™ quality level = MQ200

8.08276.0100 8.08276.0500
EM8.08276.0100CS 763 USD InStock
EM8.08276.0100 EM8.08276.0500
Vinyltriethoxysilane for synthesis, Sigma-Aldrich®
Vinyltriethoxysilane


Formula: C₈H₁₈O₃Si
MW: 190.31 g/mol
Boiling Pt: 160
Flash Pt: 34
MDL Number: MFCD00009063
CAS Number: 78-08-0
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Solubility (20 °C) (decomposition)
Molar Mass 190.31 g/mol
Boiling Point 156 °C (1013 hPa)
Vapor Pressure 7 hPa (20 °C)
Flash Point 38 °C
Refractive Index 1.3966 (25 °C, 589 nm)
Explosion Limit 0.53 - 15 %(V)
Density 0.91 g/cm3 (20 °C)
Ignition Point 265 °C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty