Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Methyl-2-chloropropionate for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Warning

M-Clarity™ quality level = MQ200

8.20331.0250 8.20331.1000
EM8.20331.0250CS 1190 USD InStock
EM8.20331.0250 EM8.20331.1000
(±)-Methyl-2-chloropropionate for synthesis, Sigma-Aldrich®
(±)-Methyl 2-chloropropionate


Formula: C₄H₇ClO₂
MDL Number: MFCD00000863
CAS Number: 17639-93-9
UN: 2933
ADR: 3,III
Order Now
Specification Test Results
Solubility (20 °C) practically insoluble
Molar Mass 122.55 g/mol
Boiling Point 127 - 129 °C
Vapor Pressure 30 hPa (20 °C)
Flash Point 36 °C DIN 51755 Part 1
Explosion Limit 2.8 - 13.9 %(V)
Density 1.13 g/cm3 (20 °C)
pH 6 - 7 (H2O, 20 °C)
Ignition Point 430 °C DIN 51794

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty