Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoprene stabilized for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP

8.21258.0100 8.21258.1000
EM8.21258.0100CS 2138 USD InStock
EM8.21258.0100 EM8.21258.1000
Isoprene stabilized for synthesis, Sigma-Aldrich®
Isoprene


Formula: C₅H₈
MDL Number: MFCD00008600
CAS Number: 78-79-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty