Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isobutylamine for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger

M-Clarity™ quality level = MQ200

8.41735.0250 8.41735.1000
EM8.41735.0250CS 634 USD InStock
EM8.41735.0250 EM8.41735.1000
Isobutylamine for synthesis, Sigma-Aldrich®
Isobutylamine


Formula: C₄H₁₁N
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: -85 °C
Density: 0.7344 g/cm³ (20 °C)
Flash Pt: -9 °C
Storage Temperature: Ambient
MDL Number: MFCD00008146
CAS Number: 78-81-9
EINECS: 201-145-4
UN: 1214
ADR: 3,II
Merck Index: 13,05149
Order Now
Specification Test Results
Solubility 1000 g/l (25 °C)
Melting Point -85 °C
Molar Mass 73.14 g/mol
Boiling Point 68 °C (1013 hPa)
Vapor Pressure 135 hPa (20 °C)
Flash Point -10 °C
Explosion Limit 2 - 9 %(V)
Density 0.736 g/cm3 (20 °C)
pH 12.2 (100 g/l, H2O, 20 °C)
Ignition Point 400 °C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty