CMUV Large Volume Metal Heating Mantles, Electrothermal
Supplier: Argos Technologies
36013-17 36013-19 36013-23
89217-416EA 4230.3 USD InStock
89217-416 89217-418 89217-424 89217-426 89217-432 89217-434
CMUV Large Volume Metal Heating Mantles, Electrothermal
Heaters Heating Mantles
CMUV Electromantles deliver the benefits of the CMU Electromantle, but do so for very large 60° funnels, as well as pear-shaped and round bottom flasks
  • Bottom opening accommodates 60° funnels of various diameters, and also accepts large capacity round bottom and pear-shaped flasks (10 - 22 L).
  • Complete heating control is provided by 3 separate electrical circuits that enable independent top, middle, and bottom heating, each controlled with its own dial switch.
  • Units ensure spill-proof protection against electric shocks via a grounded earth stainless steel screen between the flask and the heating element and double fuses.
  • The coiled heating element, suspended within a thermal insulating cartridge, provides maximum heat transfer and support.
  • Replaceable insulated heater cartridge included. Maximum element temperature is 450 °C (842 °F). Built-in energy regulator included.
  • Indicator lamps for power and heater operation.
  • The powder-coated aluminum "Cool-to-touch" outer casing is easy to clean.
  • Non-skid feet and 3 support rod clamps are also included (but not support rods)
CMUV Electromantles are available with and without controls. For controlled mantles, heating control is provided by a built-in, solid state Simmerstat energy regulator. For uncontrolled mantles, a range of external controllers are available.
This is achieved through a "V-shaped" design for the heating mantle, with funnels being accommodated through a bottom opening in the Electromantle base.
Certificaciones: CE marked.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty