Human Peripheral Blood CD14 Monocytes, Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
PB14C-1 PB14C-2 PB14C-3 PB14NC-1 PB14NC-2 PB14NC-3
101190-012EA 1650.6 USD InStock
101190-012 101190-014 101190-016 102830-074 102830-076 102830-078
Human Peripheral Blood CD14 Monocytes, Charles River Laboratories Cell Solutions
Blood
Human peripheral blood CD14 monocytes are isolated from a density gradient purified population of peripheral blood mononuclear cells (PBMCs) using either POSITIVE or NEGATIVE immunomagnetic cell separation procedures.
PBMCs are collected in HemaCare's FDA-registered collection centers following cGMP and cGTP collection guidelines from healthy human volunteer donors under IRB approved informed consent by leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol. This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugalMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty