Human Peripheral Blood M1 Macrophages, Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
PBM1C-MON-3 PBM1C-MON-1 PBM1C-MON-2
101190-028EA 3715.38 USD InStock
101190-028 101222-898 101222-900
Human Peripheral Blood M1 Macrophages, Charles River Laboratories Cell Solutions
Blood
Human peripheral blood M1 macrophages are derived from purified monocytes
PBMCs are collected in HemaCare's FDA-registered collection centers following cGMP and cGTP collection guidelines from healthy human volunteer donors under IRB approved informed consent by leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol. This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques.
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
CD14 monocytes were isolated from peripheral blood mononuclear cells (PBMCs) using positive immunomagnetic cell separation procedures and were cultured in the presence of GM-CSF for 10 days
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty