Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-COL4 Mouse Monoclonal Antibody [clone: COL-94]

Supplier: Diagnostic Biosystems
MOB229-05 MOB229
95024-418EA 360.28 USD InStock
95024-418 99990-454
Anti-COL4 Mouse Monoclonal Antibody [clone: COL-94]
Antibodies
This antibody recognizes collagen type IV epitope located on the α1 and/or α 2 chains of human collagen type IV. This antibody shows no cross-reactivity with collagen types I, II, III, V, VI and VII and does not react with human vitronectin, fibronectin or chondroitin sulfate A, B and C.

Type:
Antigen: COL4
Clonality: Monoclonal
Clone: COL-94
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty