Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-p-CK Mouse Monoclonal Antibody [clone: MNF116]

Supplier: Diagnostic Biosystems
MOB052-05 MOB052 PDM116
95024-442EA 524.51 USD InStock
95024-442 99990-216 99991-006
Anti-p-CK Mouse Monoclonal Antibody [clone: MNF116]
Antibodies
This antibody recognizes cytokeratin polypeptides ranging from 40 through 58kDa, corresponding to cytokeratins 5, 6, 8,17 and 19. It shows a broad pattern of reactivity with human epithelial tissues from simple glandular epithelia to stratified squamous epithelia, like epidermis, mammary gland ducts and tracheal epithelium.

Type:
Antigen: p-CK
Clonality: Monoclonal
Clone: MNF116
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: 0

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty