Dumont Tweezers Set in Wooden Case, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72890-01 72890-02
102098-070KT 457.19 USD InStock
102098-070 102098-072
Dumont Tweezers Set in Wooden Case, Electron Microscopy Sciences
Forceps
Dumont Tweezers Set 1: This set includes all of the fine tip Dumont tweezers made from Dumoxel steel( biological grade): specifically used in the field of microscopy for all delicate and precise work
Dumont tweezers in a fine wood display case.
Dumont Tweezers Set 1: This set includes all of the fine tip Dumont tweezers made from Dumoxel steel( biological grade): specifically used in the field of microscopy for all delicate and precise work. The kit includes one each of a Dumont style 3C, style 3, style 5, style 5/15, and style 7.
Dumont Tweezers Set 2: This set includes one each of : Style 3C epoxy coated, Style 5A, style 5/45, style 5AC, and style N5AC.
Dumont Tweezers Set 2: This set includes one each of : Style 3C epoxy coated, Style 5A, style 5/45, style 5AC, and style N5AC.
The kit includes one each of a Dumont style 3C, style 3, style 5, style 5/15, and style 7.
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty