Vial Shippers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61519-13 61519-24
102093-474PK 113.28 USD InStock
102093-474 102093-476
Vial Shippers, Electron Microscopy Sciences
Mailers and Shippers
This unique array of shipping system protects frozen and refrigerated shipping vials.
These ready-to- use kits include everything to keep temperature sensitive goods in top condition. Just load your product, add U-tek®phase change material pack, and close the shipper.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty