ThermoSafe Infectious Shipper, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61535-10 61535-60B 61535-70
102093-564PK 76.95 USD InStock
102093-564 102093-566 102093-570 102093-572
ThermoSafe Infectious Shipper, Electron Microscopy Sciences
Mailers and Shippers
ThermoSafe shippers (Model 630) include EPS foam container, flexible foam insert, clear plastic press-lock bag, secondary packaging and absorbent material for full capacity of blood, corrugated carton, and packaging instructions.
  • 4 Models – Accommodate shipments from 4 60 6 tubes / vials
  • Pre-assembled packaging includes everything needed for shipping infectious agents in accordance with DOT Regulation 49CFR Parts 171 to 178 and IATA Packaging Instructions 602.
Infectious shippers (Model INF3000) include pressure vessel with stop-lock lid, compartmentalized bubble pouch, 50 ml absorbent sheet, corrugate outer carton with required markings, shipper’s declarations and easy-to-follow instructions. Substance Shippers (model 670) protect frozen samples for over four days. The EPS foam container and lid accommodates either model 630 or model 660B shipper plus 25 pounds of dry ice. Foam insert holds in place while dry ice sublimates. Includes one class 9 miscellaneous dangerous goods label (for dry ice) and shipping carton (dry ice NOT included). Note: Compliance with UN/DOT regulations governing shipments of infectious substances in these approved shippers is contingent on the use of all supplied components. The shipper is ultimately responsible for determining the suitability of packaging for use with contents. Consult appropriate regulations.
Certificaciones: Meet US Dept of Transportation (DOT) and IATA requirement for transporting infectious substances (UN2814 and UN 2900).
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty