ThermoSafe Medical Transporter, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61538 61537-31 61537-32 61537-40 61537-41
102091-050EA 455.79 USD InStock
102091-050 102093-606 102093-608 102093-610 102093-612
ThermoSafe Medical Transporter, Electron Microscopy Sciences
Mailers and Shippers
Model 930
Insulated expanded polypropylene medical transporter is ideal for on-site collection, storage, and transport of temperature sensitive specimens, medications, medical devices and organs. Durable nylon clad transporter with impact resistant expanded polyethylene container.
Model 930
For operating rooms storage and blood bank transport, provides temperature control for packed red blood cells and other temperature sensitive specimens
Qualified by ISC labs with three 32oz Polar Pack Rigid Bottle refrigerant and two 16oz Polar Pack refrigerant.
Maintains up to 8 packed red blood cell units at 1°C – 6°C while in storage for 6 hours or more and between 1°C – 10°C while in transit for up to 24 hours.
Including operating instruction and test data for storage of packed cells.
Model 960
For mobile blood collection, cools and stores whole blood units and other temperature sensitive materials
Qualified by ISC labs for temporary storage and transport of both platelet (20°C – 24°C for up to 8 hours) and non-platelet production (1°C – 10°C for up to 24 hours) whole blood with crushed wet ice.
Includes operating instructions and test data for cooling platelet and non- platelet production blood.
Volume: 1.60 cu. Ft. and 1 wall thickness
Model 931, 932
Includes removable, rigid, thermoformed tray – ideal for washing and reusing; prevents leaking inside and out of transport unit, reinforced durability. Adjustable, durable carrying strap for shoulder transport. Model 932 also features a comfortable sturdy hand strap.
Model 931: 0.3 cu. ft. volume and 3/8 wall thickness
Model 932: 0.53 cu. ft. volume and 3/8 wall thickness
Model 640, 641
Inner partition divides unit and separates specimens. Features built-in lid compartments for U-tek® refrigerant packs. Adjustable nylon carrying strap for hand or shoulder. Model 641 includes a vial foam insert that holds twenty four 5 - 15 mL vials and one model 418 U-tek® Volume is 0.30 cu. Ft. with1/4 wall thickness.
Model 960
For mobile blood collection, cools and stores whole blood units and other temperature sensitive materials
Qualified by ISC labs for temporary storage and transport of both platelet (20°C – 24°C for up to 8 hours) and non-platelet production (1°C – 10°C for up to 24 hours) whole blood with crushed wet ice.
Includes operating instructions and test data for cooling platelet and non- platelet production blood.
Volume: 1.60 cu. Ft. and 1 wall thickness
Model 931, 932
Includes removable, rigid, thermoformed tray – ideal for washing and reusing; prevents leaking inside and out of transport unit, reinforced durability. Adjustable, durable carrying strap for shoulder transport. Model 932 also features a comfortable sturdy hand strap.
Model 931: 0.3 cu. ft. volume and 3/8 wall thickness
Model 932: 0.53 cu. ft. volume and 3/8 wall thickness
Model 640, 641
Inner partition divides unit and separates specimens. Features built-in lid compartments for U-tek® refrigerant packs. Adjustable nylon carrying strap for hand or shoulder. Model 641 includes a vial foam insert that holds twenty four 5 - 15 mL vials and one model 418 U-tek® Volume is 0.30 cu. Ft. with1/4 wall thickness.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty