Geological Thin Section Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75941-15 75941-15H
102681-248EA 393.07 USD InStock
102681-248 102681-250
Geological Thin Section Holder, Electron Microscopy Sciences
Slide Holders
This thin section holder is made from aluminum with copper clips and brass screws.
  • Holder has convenient tweezer inserts
  • Allows for easy loading
The unit measures 55 x 30 x 8 mm and holds a standard geological thin section or slide measuring 47 x 27 mm. The holder comes complete with 2 clips to insure nothing moves.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty