Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-GYPA Mouse Monoclonal Antibody [clone: JC159]

Supplier: Diagnostic Biosystems
MOB066-05 PDM124 MOB066
95024-566EA 435.76 USD InStock
95024-566 99991-020 99990-242
Anti-GYPA Mouse Monoclonal Antibody [clone: JC159]
Antibodies
This antibody is specific to erythroid cells at various stages of differentiation from the erythroblast to the mature red cell.

Type:
Antigen: Glycophorin A
Clonality: Monoclonal
Clone: JC159
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty