Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-HAS Mouse Monoclonal Antibody [clone: OCH1E5]

Supplier: Diagnostic Biosystems
MOB426-05 MOB426 PDM162
95024-584EA 439.74 USD InStock
95024-584 99990-786 10064-976
Anti-HAS Mouse Monoclonal Antibody [clone: OCH1E5]
Antibodies
This antibody recognizes an uncharacterized antigen present in both adults and fetal normal hepatocytes to produce a distinct granular cytoplasmic staining. This antibody stains majority of hepatocellular carcinomas.

Type:
Antigen: Hepatocyte Specific Antigen
Clonality: Monoclonal
Clone: OCH1E5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty