Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-p504S/α-Methylacyl-CoA Racemase (AMACR) Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems
RP134-05 PDR046 RP134
95025-110EA 419.89 USD InStock
95025-110 99991-122 99991-434
Anti-p504S/α-Methylacyl-CoA Racemase (AMACR) Rabbit Polyclonal Antibody
Antibodies
P504S has been recently described as a prostate cancer-specific gene that encodes a protein involved in the β-oxidation of branched chain fatty acids. Expression of p504S protein is found in prostatic adenocarcinoma but not in benign prostatic tissue. It stains premalignant lesions of prostate: high-grade prostatic intraepithelial neoplasia (PIN) and atypical adenomatous hyperplasia. P504S can be used as a positive marker for PIN.

Type:
Antigen: AMCAR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty