Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-MGMT Mouse Monoclonal Antibody [clone: MT3.1]

Supplier: Diagnostic Biosystems
MOB423-05 MOB423
95024-676EA 576.15 USD InStock
95024-676 99990-782
Anti-MGMT Mouse Monoclonal Antibody [clone: MT3.1]
Antibodies
O(6)-methylguanine-DNA-methyltransferase (MGMT) is a 22 kDa human DNA repair protein that removes O(6)-alkylguanine DNA adducts. MGMT acts as a suicide acceptor protein repairing the DNA by accepting alkyl groups and consequently inactivating itself. This antibody stains all mantle zone lymphocytes and 50% of germinal center lymphocytes. Basaloid epithelial cells of tonsil squamous mucosa are also stained positive with this antibody.

Type:
Antigen: MGMT
Clonality: Monoclonal
Clone: MT3.1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty