Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Thyroglobulin Mouse Monoclonal Antibody [clone: 1D4]

Supplier: Diagnostic Biosystems
MOB223 MOB223-05
99990-444EA 584.13 USD InStock
99990-444 95024-838
Anti-Thyroglobulin Mouse Monoclonal Antibody [clone: 1D4]
Antibodies
This antibody recognizes thyroglobulin in hyperplastic and neoplastic thyroid. Thyroglobulin is a protein of 670 kDa consisting of two identical subunits. Thyroglobulin is synthesized by the follicular epithelial cells of the thyroid.

Type:
Antigen: Thyroglobulin
Clonality: Monoclonal
Clone: 1D4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty