Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-TGM2 Mouse Monoclonal Antibody [clone: CUB7402]

Supplier: Diagnostic Biosystems
MOB353-05 MOB353
95024-860EA 572.2 USD InStock
95024-860 99990-660
Anti-TGM2 Mouse Monoclonal Antibody [clone: CUB7402]
Antibodies
This antibody reacts with a 77-85 kDa protein known as transglutaminase II (TGase II). TGase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. Different tissue and cell types express varying amounts of tissue transglutaminase.

Type:
Antigen: TGM2
Clonality: Monoclonal
Clone: CUB7402
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty