Spray Nine® Heavy Duty Cleaner/Degreaser/Disinfectant
Supplier: Janitorial Supplies
ITW26825 ITW26832CT
10048-974CS 229.8 USD InStock
10048-974 10048-976
Spray Nine® Heavy Duty Cleaner/Degreaser/Disinfectant
Cleaning Agents Disinfectants
A professional-grade cleaner and disinfectant that removes all types of dirt, grease, grime and stains.
  • Cleans, degreases and deodorizes
  • Disinfects against viruses in 30 seconds
  • Disinfects against bacteria in 45 seconds
  • Sanitizes in 10 seconds
  • Controls mold and mildew
  • Removes stains
Kills fungus in three minutes. Effective against dirt, oil, grease, hydraulic fluid, exhaust residue, fresh paint, wallpaper paste, soap scum, burned-on food, soot, ash, smoke film, carpet stains, fabric stains, scuff marks, bacteria, dust, fungus, germs, grime, mildew, mold, organic matter and viruses.
For use in bathrooms, breakrooms, kitchens, offices, manufacturing plants, appliances, equipment, automotive, medical offices and exam rooms, hospitals/healthcare, veterinary medicine, machinery/tools. For use on tables, walls, floors, countertops, exterior surfaces of refrigerators, Tubs, showers, glazed tile, toilet exteriors, garbage pails.
Physical Specifications: N-Alkyl Dimethyl Benzyl Ammonium Chloride; N-Alkyl Dimethyl Ethybenzyl Ammonium Chloride; Ethoxylated Alcohols; Dipropylene Glycol Mononbutyl Ether.
Certificaciones: NSF/USDA Classification C1.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty