Rota-Rack® Adjustable Tube Racks
Supplier: Heathrow Scientific
HS29040A HS29040B
490012-900EA 105.72 USD InStock
490012-900 490012-902
Rota-Rack® Adjustable Tube Racks
Racks Tube Racks
Each module is interlocked and adjustable with just a twist allowing to work with multiple sized tubes in the same rack
  • Made from polypropylene
  • Suitable for a massive variety of culture, centrifuge, micro centrifuge, round, flat or conical bottomed tubes
Each module of the large version holds two 50 ml tubes, eight 15 ml tubes, six 20 mm tubes, or ten 10 to 13 mm tubes.
Each module of the small version holds six 15 ml tubes, nine 1.5/2.0 ml tubes, twelve 0.5/0.6 ml tubes, or thirty-two independent 0.2 ml PCR tubes or four 8-tube strips.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty