Vinyl-Coated Lead Rings
Supplier: Heathrow Scientific
HS8881A HS8881B HS8881C HS8882B HS8882C HS8882D HS8882A
490007-654CS 4383.11 USD InStock
490007-654 490007-656 490007-658 490007-660 490007-662 490007-664 490012-176
Vinyl-Coated Lead Rings
Weights Flask Weights
Vinyl Coated as Not to Scratch Bench Tops or Glassware.
  • Designed to stabilize glassware and plasticware immersed in waterbaths
C-shape format allows some manual size variation for a customized fit.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty