Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydrochloric acid 37% ACS

Supplier: RICCA CHEMICAL CO
Danger

RABH0010-2.5D1 RABH0010-4C
RCRABH001025D1EA 144 USD InStock
RCRABH001025D1 RCRABH0010-4C RCRABH0010-55E
Hydrochloric acid 37% ACS
Hydrochloric acid
Testing reagent for ethanol plants.


Formula: HCl
MW: 36.46
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 II
Order Now
Specification Test Results
Ammonium (NH₄) max 3 ppm
Arsenic (As) max 0.01 ppm
Assay 36.5-38.0 % HCl
Bromide (Br) max 0.005 %
Color (APHA) max 10
Free Chlorine (Cl) max 1 ppm
Heavy Metals (as Pb) max 1 ppm
Iron (Fe) max 0.2 ppm
Residue after Ignition max 5 ppm
Sulfate (SO₄) max 1 ppm
Sulfite (SO₃) max 1 ppm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty