Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-HAUS8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
109-401-C39S 109-401-C39
RL109-401-C39SEA 151.49 USD InStock
RL109-401-C39S RL109-401-C39
Anti-HAUS8 Rabbit Polyclonal Antibody
Antibodies
This antiserum has been tested for use in ELISA and western blotting using a full length recombinant Hice1 Protein. Specific conditions for reactivity and detection of Hice1 should be optimized by the end user. Expect a band approximately ~45 kDa in size.

Recommended Dilutions: ELISA: 1:250000; Western Blot: 1:10000;

Type: Primary
Antigen: HICE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty