Human Recombinant IL1 beta (from E. coli)
Supplier: PROSCI INCORPORATED
40-101 40-110 40-241
10072-018EA 305 USD InStock
10072-018 10072-022 10072-360
Human Recombinant IL1 beta (from E. coli)
Proteins and Peptides
IL-1beta is a proinflammatory cytokine produced in a variety of cells including monocytes, tissue macrophages, keratinocytes and other epithelial cells. Both IL-1alpha and IL-1beta binds to the same receptor and has similar if not identical biological properties. These cytokines have a broad range of activities including, stimulation of thymocyte proliferation, by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF-like activity and the ability to stimulate the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1beta is a secreted cytokine, IL-1alpha is predominantly a cell-associated cytokine. Recombinant rat IL-1beta is a 17.3 kDa protein containing 153 amino acid residues. Recombinant murine IL-1beta is a 17.5 kDa protein containing 153 amino acid residues. Recombinant human IL-1beta is a 17.3 kDa protein containing 153 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty