Rat recombinant IL10 (from E.coli)
Supplier: PROSCI INCORPORATED
40-417 40-471 40-253
10072-400EA 305 USD InStock
10072-400 10072-492 10072-830
Rat recombinant IL10 (from E.coli)
Proteins and Peptides
IL-10 is an immunosuppressive cytokine produced by a variety of mammalian cell types including macrophages, monocytes, T cells, B cells and keratinocytes. IL-10 inhibits the expression of proinflammatory cytokines such as IL-1 and TNFα. Like IL-4, IL-10 enhances humoral immune responses and attenuates cell-mediated immune reactions. Human IL-10 active on murine cells, but murine IL-10 is inactive on human cells. Recombinant murine IL-10 is an 18.7 kDa protein of 161 amino acid residues. Recombinant rat IL-10 is an 18.7 kDa protein of 160 amino acid residues. Recombinant human IL-10 is an 18.6 kDa protein of 161 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty