Rat Recombinant IL7 (from E. coli)
Supplier: PROSCI INCORPORATED
40-251 40-561 40-435 40-467
10072-066EA 305 USD InStock
10072-066 10072-188 10072-430 10072-484
Rat Recombinant IL7 (from E. coli)
Proteins and Peptides
IL-7 is a hematopoietic growth factor, which affects primarily early B and T cells. Produced by thymic stromal cells, spleen cells and keratinocytes, IL-7 can also co-stimulate the proliferation of mature T cells in combination with other factors such as ConA and IL-2. Human and murine IL-7 is cross-species reactive. Recombinant human IL-7 is a 17.4 kDa protein containing 153 amino acid residues. Recombinant rat IL-7 is a 15.0 kDa protein containing 130 amino acid residues. Recombinant murine IL-7 is a 15.0 kDa protein containing 130 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty