Rat Recombinant Prolactin (from E. coli)
Supplier: PROSCI INCORPORATED
40-456 40-478 40-152
10072-466EA 305 USD InStock
10072-466 10072-506 10072-688
Rat Recombinant Prolactin (from E. coli)
Proteins and Peptides
Prolactin is a neuroendocrine hormone secreted by the pituitary gland. Its primary function is to promote and maintain lactation during pregnancy and suckling. In addition, Prolactin plays an immune-regulatory role by stimulating the activities of ornithine decarboxylase and protein kinase C, which are important for the proliferation, differentiation, and function of lymphocytes. Recombinant human Prolactin is a 23 kDa globular protein containing 200 amino acid residues. Recombinant murine Prolactin is a 22.5 kDa globular protein containing 198 amino acid residues. Recombinant Rat Prolactin is a 22.6 kDa globular protein containing 198 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty