Mouse Recombinant FGF9 (from E. coli)
Supplier: PROSCI INCORPORATED
40-489 40-170
10072-522EA 305 USD InStock
10072-522 10072-706
Mouse Recombinant FGF9 (from E. coli)
Proteins and Peptides
FGF-9 is a heparin binding growth factor that belongs to the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-9 targets glial cells, astrocytes cells and other cells that express the FGFR 1c, 2c, 3b, 3c, and 4. Recombinant human FGF-9 is a 23.4 kDa protein consisting of 207 amino acid residues. Recombinant murine FGF-9 is a 23.3 kDa protein containing 205 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty