Human Recombinant KGF (from E. coli)
Supplier: PROSCI INCORPORATED
40-161 40-605
10072-042EA 305 USD InStock
10072-042 10073-094
Human Recombinant KGF (from E. coli)
Proteins and Peptides
Keratinocyte Growth Factor (KGF/FGF-7) is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of variety of tissues, by promoting cellular proliferation and differentiation. KGF/FG-7 is a mitogen factor specific for epithelial cells and keratinocytes and signals through FGFR 2b. KGF/FGF-7 plays a role in kidney and lung development, angiogenesis, and wound healing. Recombinant human KGF/FGF-7 is an 18.9 kDa protein consisting of 163 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty