Human Recombinant NT3 (from E. coli)
Supplier: PROSCI INCORPORATED
40-497 40-565
10072-228EA 305 USD InStock
10072-228 10073-038
Human Recombinant NT3 (from E. coli)
Proteins and Peptides
NT-3 is a neurotrophic factor structurally related to β-NGF, BDNF, and NT-4. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. NT-3 is expressed by neurons of the central nervous systems and can signal through the trk receptors. NT-3 promotes the growth and survival of nerve and glial cells. The amino acid sequences of human, murine and rat NT-3 are identical. Recombinant human NT-3 is a noncovalently linked homodimer, of two 13.6 kDa polypeptide monomers (240 total amino acid residues). Human and Mouse NT-3 sequences are identical.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty