Human Recombinant VEGF121 (from E. coli)
Supplier: PROSCI INCORPORATED
40-163 40-612
10072-046EA 305 USD InStock
10072-046 10073-106
Human Recombinant VEGF121 (from E. coli)
Proteins and Peptides
VEGF is a potent growth and angiogenic cytokine. It stimulates proliferation and survival of endothelial cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays a prominent role in normal and pathological angiogenesis. VEGF signals through three receptors; fms-like tyrosine kinase (flt-1), KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Due to its increased acidity, VEGF121 circulates more freely than other VEGF forms, which bind more tightly with vascular heparin sulfates. Recombinant human VEGF121 is a 28.4 kDa disulfide-linked homodimeric protein consisting of two 121 amino acid polypeptide chains.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty