STAR™ DISH Petri Dishes, Nesting Alpine, Phoenix
Supplier: Phoenix Biomedical
321 361 360
25389-012CS 313.82 USD InStock
25389-012 25389-028 25389-026
STAR™ DISH Petri Dishes, Nesting Alpine, Phoenix
Culture Dishes Petri Dishes
The unique recessed lid on these dishes offers excellent stacking stability and saves space
The flat bottom allows a reduction in the amount of media required. Patented cross ribs on the bottom make these dishes resilient to warpage when media is poured at a high temperature. These dishes are excellent for performing colony counts because the cross ribs also provide a useful border for making quadrents.
They work with most models of media pouring equipment. Available with four ventilation plugs on the underside of the lid allow for venting and prevent condensation from developing and dripping onto the culture.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty