STAR™ DISH High Profile' Petri Dishes, Phoenix Biomedical'
Supplier: Phoenix Biomedical
002 307
25389-000CS 304.45 USD InStock
25389-000 89031-138
STAR™ DISH High Profile' Petri Dishes, Phoenix Biomedical'
Culture Dishes Petri Dishes
These high-profile petri dishes are specifically designed to accommodate more media
They are excellent for mycology, histology and pathology labs, as well as seed germination work. These dishes are ideal when longer culture periods are required.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty