Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulphuric acid 72% (w/w)

Supplier: RICCA CHEMICAL CO

R8191600-1A R8191600-4A
RCR8191600-1AEA 86 USD InStock
RCR8191600-1A RCR8191600-4A
Sulphuric acid 72% (w/w)
Sulfuric acid


Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8 II
Order Now
Specification Test Results
Assay 72 ± 0.2 % (w/w)
Appearance Colorless liquid

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty