KWIK-STIK Candida tropicalis ATCC® 13803™, Microbiologics
Supplier: Ofite
0450K 0450P
89504-426EA 303.35 USD InStock
89504-426 89504-430
KWIK-STIK Candida tropicalis ATCC® 13803™, Microbiologics
Controls and Calibration Verification Materials
The KWIK-STIK™ is well-known for its clever design and accurate, repeatable results
Each KWIK-STIK™ device features a single QC microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid, and an inoculating swab. For added convenience, a peel-off identification label is included for easy documentation.
Certificaciones: The KWIK-STIK™ falls under the category of In-Vitro Diagnostics Medical Devices Directives for CE Mark Conformity. This product is also listed with the FDA.
Información para pedidos: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty